Assay Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118853.2 | XP_014974339.1 | 1041 | 67 | ||
| XM_015118923.2 | XP_014974409.1 | 782 | 67 | |||
| GenBank mRNA | JU329170.1 | - | 422 | 67 | ||
| JV723612.1 | - | 514 | 67 | |||
More Information
Set Membership: |
|

AGMAT
Genomic Map