Assay Details
Gene Symbol: BICD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151407.2 | XP_015006893.1 | 2765 | 69 | ||
XM_015151408.2 | XP_015006894.1 | 2765 | 69 | |||
XM_015151409.2 | XP_015006895.1 | 2765 | 69 | |||
XM_015151410.2 | XP_015006896.1 | 2877 | 69 | |||
XM_028829397.1 | XP_028685230.1 | 2248 | 69 | |||
XM_028829398.1 | XP_028685231.1 | 2248 | 69 | |||
GenBank mRNA | JU328343.1 | - | 1281 | 69 | ||
JV489171.1 | - | 2253 | 69 | |||
JV720836.1 | - | 2331 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
