Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149658.2 | XP_015005144.2 | 1698 | 103 | ||
| XM_015149659.2 | XP_015005145.2 | 1601 | 103 | |||
| GenBank mRNA | JU475991.1 | - | 1491 | 103 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map