Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149268.2 | XP_015004754.2 | 2501 | 70 | ||
GenBank mRNA | JU342559.1 | - | 122 | 70 | ||
JU470964.1 | - | 1916 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
