Assay Details
Gene Symbol: LOC713572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143401.2 | XP_014998887.1 | 512 | 87 | ||
XM_015143402.2 | XP_014998888.1 | 565 | 87 | |||
XM_028851425.1 | XP_028707258.1 | 154 | 87 | |||
GenBank mRNA | JU319755.1 | - | 184 | 87 | ||
JV043570.1 | - | 178 | 87 | |||
JV636544.1 | - | 192 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015143401.2, XM_015143402.2, XM_028851425.1 transcripts.
Set Membership: |
![]() ![]() |
