Assay Details
| Gene Symbol: MRPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112931.2 | XP_014968417.1 | 805 | 83 | ||
| XM_015112934.2 | XP_014968420.1 | 604 | 83 | |||
| XM_015112936.2 | XP_014968422.1 | 630 | 83 | |||
| XM_015112937.2 | XP_014968423.1 | 411 | 83 | |||
| XM_028832228.1 | XP_028688061.1 | 411 | 83 | |||
| GenBank mRNA | JU334840.1 | - | 470 | 83 | ||
| JU334841.1 | - | 470 | 83 | |||
| JU475867.1 | - | 453 | 83 | |||
| JV047270.1 | - | 521 | 83 | |||
| JV527805.1 | - | 432 | 83 | |||
| JV716418.1 | - | 499 | 83 | |||
More Information
Set Membership: |
|

MRPL30
Genomic Map