Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112153.2 | XP_014967639.1 | 263 | 94 | ||
XM_015112154.2 | XP_014967640.1 | 306 | 94 | |||
GenBank mRNA | JU332214.1 | - | 139 | 94 | ||
JV661739.1 | - | 201 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
