Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084635.4 | XP_001084635.2 | 2887 | 75 | ||
XM_015118154.2 | XP_014973640.1 | 2939 | 75 | |||
GenBank mRNA | JU325912.1 | - | 2614 | 75 | ||
JU475783.1 | - | 2780 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
