Assay Details
| Gene Symbol: FZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146845.2 | XP_015002331.2 | 1884 | 81 | ||
| GenBank mRNA | JU331971.1 | - | 627 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FZD8| Gene Symbol: FZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146845.2 | XP_015002331.2 | 1884 | 81 | ||
| GenBank mRNA | JU331971.1 | - | 627 | 81 | ||
Set Membership: |
|