Assay Details
| Gene Symbol: COX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147913.2 | XP_015003399.2 | 3595 | 103 | ||
| GenBank mRNA | JU338952.1 | - | 623 | 103 | ||
| JV707191.1 | - | 623 | 103 | |||
More Information
Set Membership: |
|

COX15
Genomic Map