Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134378.2 | XP_014989864.2 | 776 | 90 | ||
| GenBank mRNA | JU336662.1 | - | 746 | 90 | ||
| JU477462.1 | - | 761 | 90 | |||
| JV048418.1 | - | 758 | 90 | |||
| JV663184.1 | - | 754 | 90 | |||
More Information
Set Membership: |
|

ZNF394
Genomic Map