Assay Details
Gene Symbol: CEP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109724.4 | XP_001109724.3 | 101 | 95 | ||
XM_015125697.2 | XP_014981183.1 | 101 | 95 | |||
XM_015125698.2 | XP_014981184.1 | 101 | 95 | |||
GenBank mRNA | JU474274.1 | - | 39 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109724.4, XM_015125697.2, XM_015125698.2 transcripts.
Set Membership: |
![]() ![]() |
