Assay Details
Gene Symbol: LOC114669852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842799.1 | XP_028698632.1 | 992 | 78 | ||
GenBank mRNA | JU476522.1 | - | 958 | 78 | ||
JV066501.1 | - | 971 | 78 |
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124582.2 | XP_014980068.2 | 1159 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
