Assay Details
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150935.2 | XP_015006421.1 | 150 | 71 | ||
GenBank mRNA | JU495045.1 | - | 81 | 71 | ||
JV073687.1 | - | 54 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015150935.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
