Assay Details
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122966.2 | XP_014978452.2 | 600 | 59 | ||
XM_028838688.1 | XP_028694521.1 | 394 | 59 | |||
GenBank mRNA | JV046180.1 | - | 518 | 59 | ||
JV635952.1 | - | 501 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
