Assay Details
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122727.2 | XP_014978213.1 | 505 | 91 | ||
XM_028839189.1 | XP_028695022.1 | 805 | 91 | |||
GenBank mRNA | JU325236.1 | - | 78 | 91 | ||
JV062624.1 | - | 485 | 91 | |||
JV653028.1 | - | 483 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
