Assay Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143697.2 | XP_014999183.1 | 726 | 71 | ||
| GenBank mRNA | JU327030.1 | - | 175 | 71 | ||
| JV613780.1 | - | 318 | 71 | |||
| JV712531.1 | - | 291 | 71 | |||
More Information
Set Membership: |
|

GPR137C
Genomic Map