Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149162.2 | XP_015004648.2 | 1106 | 99 | ||
| XM_015149220.2 | XP_015004706.2 | 457 | 99 | |||
| XM_028840044.1 | XP_028695877.1 | 692 | 99 | |||
| XM_028840055.1 | XP_028695888.1 | 721 | 99 | |||
| GenBank mRNA | JU322922.1 | - | 350 | 99 | ||
| JU332336.1 | - | 350 | 99 | |||
| JU472384.1 | - | 364 | 99 | |||
| JV045793.1 | - | 410 | 99 | |||
| JV635870.1 | - | 402 | 99 | |||
More Information
Set Membership: |
|

NMNAT1
Genomic Map