Assay Details
| Gene Symbol: PLCXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126854.2 | XP_014982340.2 | 471 | 73 | ||
| XM_015126855.2 | XP_014982341.2 | 1559 | 73 | |||
| XM_015126856.2 | XP_014982342.2 | 686 | 73 | |||
| XM_015126857.2 | XP_014982343.2 | 1876 | 73 | |||
| XM_028842027.1 | XP_028697860.1 | 662 | 73 | |||
| XM_028842028.1 | XP_028697861.1 | 432 | 73 | |||
| GenBank mRNA | JU335323.1 | - | 411 | 73 | ||
| JU473305.1 | - | 423 | 73 | |||
| JV047561.1 | - | 416 | 73 | |||
| JV655407.1 | - | 422 | 73 | |||
More Information
Set Membership: |
|

PLCXD1
Genomic Map