Assay Details
| Gene Symbol: ZYG11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139848.2 | XP_014995334.2 | 1620 | 60 | ||
| XM_028833737.1 | XP_028689570.1 | 1296 | 60 | |||
| GenBank mRNA | JV651981.1 | - | 276 | 60 | ||
More Information
Set Membership: |
|

ZYG11A
Genomic Map