Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113460.4 | XP_001113460.1 | 415 | 66 | ||
XM_028835036.1 | XP_028690869.1 | 415 | 66 | |||
GenBank mRNA | JU497098.1 | - | 205 | 66 | ||
JV666809.1 | - | 205 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
