Assay Details
| Gene Symbol: B3GALT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108171.4 | XP_001108171.2 | 2061 | 61 | ||
| GenBank mRNA | JV705114.1 | - | 616 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3GALT5| Gene Symbol: B3GALT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108171.4 | XP_001108171.2 | 2061 | 61 | ||
| GenBank mRNA | JV705114.1 | - | 616 | 61 | ||
Set Membership: |
|