Assay Details
Gene Symbol: ABI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119336.2 | XP_014974822.2 | 1155 | 62 | ||
XM_015119338.2 | XP_014974824.2 | 1433 | 62 | |||
GenBank mRNA | JU325434.1 | - | 123 | 62 | ||
JU487745.1 | - | 129 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
