Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124803.2 | XP_014980289.2 | 637 | 91 | ||
GenBank mRNA | JV692211.1 | - | 474 | 91 | ||
KT749897.1 | - | 367 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
