Assay Details
| Gene Symbol: MRPS31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120961.2 | XP_014976447.1 | 1030 | 71 | ||
| XM_015120962.2 | XP_014976448.2 | 836 | 71 | |||
| XM_015120963.2 | XP_014976449.1 | 931 | 71 | |||
| GenBank mRNA | JU332456.1 | - | 868 | 71 | ||
| JU473923.1 | - | 857 | 71 | |||
| JV045871.1 | - | 791 | 71 | |||
| JV639944.1 | - | 861 | 71 | |||
| JV705034.1 | - | 858 | 71 | |||
More Information
Set Membership: |
|

MRPS31
Genomic Map