Assay Details
Gene Symbol: LOC708234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028853105.1 | XP_028708938.1 | 1142 | 104 | ||
GenBank mRNA | JU326537.1 | - | 764 | 104 |
Gene Symbol: LOC114669859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028853162.1 | XP_028708995.1 | 1233 | 104 | ||
XM_028853163.1 | XP_028708996.1 | 1142 | 104 | |||
GenBank mRNA | JV614347.1 | - | 1054 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
