Assay Details
Gene Symbol: LOC707412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126190.2 | XP_014981676.2 | 93 | |||
XM_015126191.2 | XP_014981677.2 | 93 | ||||
XM_028841585.1 | XP_028697418.1 | 93 | ||||
GenBank mRNA | JU332570.1 | - | 369 | 93 | ||
JU332571.1 | - | 369 | 93 | |||
JV454629.1 | - | 214 | 93 | |||
JV565056.1 | - | 348 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
