Assay Details
| Gene Symbol: LOC695175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831976.1 | XP_028687809.1 | 2839 | 56 | ||
| GenBank mRNA | JV573593.1 | - | 617 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC695175| Gene Symbol: LOC695175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831976.1 | XP_028687809.1 | 2839 | 56 | ||
| GenBank mRNA | JV573593.1 | - | 617 | 56 | ||
Set Membership: |
|