Assay Details
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141529.2 | XP_014997015.1 | 3944 | 77 | ||
XM_015141530.2 | XP_014997016.1 | 4594 | 77 | |||
GenBank mRNA | JV723762.1 | - | 1317 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
