Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123640.2 | XP_014979126.2 | 986 | 94 | ||
XM_015123643.2 | XP_014979129.2 | 647 | 94 | |||
GenBank mRNA | JU322954.1 | - | 113 | 94 | ||
JV544581.1 | - | 113 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123640.2, XM_015123643.2 transcripts.
Set Membership: |
![]() ![]() |
