Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123640.2 | XP_014979126.2 | 1257 | 83 | ||
XM_015123641.2 | XP_014979127.2 | 1140 | 83 | |||
XM_015123643.2 | XP_014979129.2 | 918 | 83 | |||
XM_028839056.1 | XP_028694889.1 | 596 | 83 | |||
XM_028839057.1 | XP_028694890.1 | 801 | 83 | |||
XM_028839058.1 | XP_028694891.1 | 695 | 83 | |||
XM_028839059.1 | XP_028694892.1 | 523 | 83 | |||
XM_028839060.1 | XP_028694893.1 | 504 | 83 | |||
XM_028839061.1 | XP_028694894.1 | 1095 | 83 | |||
GenBank mRNA | JU322954.1 | - | 384 | 83 | ||
JV544581.1 | - | 384 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
