Assay Details
| Gene Symbol: PRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122741.2 | XP_014978227.2 | 3433 | 70 | ||
| XM_028839525.1 | XP_028695358.1 | 3433 | 70 | |||
| GenBank mRNA | JV073105.1 | - | 134 | 70 | ||
More Information
Set Membership: |
|

PRAM1
Genomic Map