Assay Details
| Gene Symbol: LOC100423439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124999.2 | XP_014980485.2 | 2199 | 122 | ||
| XM_015125000.2 | XP_014980486.2 | 2845 | 122 | |||
| GenBank mRNA | JU341467.1 | - | 218 | 122 | ||
More Information
Set Membership: |
|

LOC100423439
Genomic Map