Assay Details
| Gene Symbol: NRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143427.2 | XP_014998913.2 | 523 | 62 | ||
| XM_028851439.1 | XP_028707272.1 | 403 | 62 | |||
| GenBank mRNA | JU475748.1 | - | 93 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015143427.2, XM_028851439.1 transcripts.
Set Membership: |
|

NRL
Genomic Map