Assay Details
Gene Symbol: NUTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126441.2 | XP_014981927.1 | 642 | 115 | ||
XM_015126442.2 | XP_014981928.1 | 381 | 115 | |||
XM_028841534.1 | XP_028697367.1 | 448 | 115 | |||
XM_028841535.1 | XP_028697368.1 | 285 | 115 | |||
GenBank mRNA | JU320964.1 | - | 265 | 115 | ||
JU320965.1 | - | 265 | 115 | |||
JU332446.1 | - | 265 | 115 | |||
JU473515.1 | - | 226 | 115 | |||
JV044952.1 | - | 275 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
