Assay Details
Gene Symbol: FAM200B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099745.4 | XP_001099745.2 | 466 | 114 | ||
XM_015138076.2 | XP_014993562.2 | 469 | 114 | |||
XM_015138077.2 | XP_014993563.2 | 452 | 114 | |||
XM_015138078.2 | XP_014993564.2 | 510 | 114 | |||
XM_015138079.2 | XP_014993565.2 | 453 | 114 | |||
XM_015138081.2 | XP_014993567.2 | 439 | 114 | |||
XM_015138082.2 | XP_014993568.2 | 493 | 114 | |||
XM_015138083.2 | XP_014993569.2 | 375 | 114 | |||
XM_028848362.1 | XP_028704195.1 | 466 | 114 | |||
GenBank mRNA | JU484952.1 | - | 325 | 114 | ||
JV649054.1 | - | 325 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001099745.4, XM_015138076.2, XM_015138077.2, XM_015138078.2, XM_015138079.2, XM_015138081.2, XM_015138082.2, XM_015138083.2, XM_028848362.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
