Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092994.3 | XP_001092994.2 | 506 | 70 | ||
| GenBank mRNA | JU483710.1 | - | 126 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAFF| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092994.3 | XP_001092994.2 | 506 | 70 | ||
| GenBank mRNA | JU483710.1 | - | 126 | 70 | ||
Set Membership: |
|