Assay Details
Gene Symbol: PPP1R15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097695.4 | XP_001097695.2 | 2841 | 78 | ||
GenBank mRNA | JU336791.1 | - | 2222 | 78 | ||
JU477551.1 | - | 2220 | 78 | |||
JV663862.1 | - | 2229 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
