Assay Details
Gene Symbol: GADD45GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123048.2 | XP_014978534.2 | 449 | 83 | ||
GenBank mRNA | JU325499.1 | - | 321 | 83 | ||
JV667286.1 | - | 410 | 83 | |||
JV726467.1 | - | 334 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
