Assay Details
| Gene Symbol: NR2C2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123426.2 | XP_014978912.1 | 1150 | 81 | ||
| GenBank mRNA | JU332904.1 | - | 1124 | 81 | ||
| JU474297.1 | - | 1013 | 81 | |||
| JV051593.1 | - | 340 | 81 | |||
More Information
Set Membership: |
|

NR2C2AP
Genomic Map