Assay Details
Gene Symbol: LOC706967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832145.1 | XP_028687978.1 | 1382 | 83 | ||
GenBank mRNA | JU334757.1 | - | 1215 | 83 |
Gene Symbol: LOC704637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808045.3 | XP_002808091.3 | 1233 | 83 | ||
XM_028832146.1 | XP_028687979.1 | 1068 | 83 | |||
GenBank mRNA | JU334758.1 | - | 1208 | 83 | ||
JU475799.1 | - | 1229 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
