Assay Details
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148700.2 | XP_015004186.1 | 2565 | 83 | ||
GenBank mRNA | JU322244.1 | - | 2419 | 83 | ||
JU471252.1 | - | 2411 | 83 | |||
JV639096.1 | - | 2343 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
