Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125024.2 | XP_014980510.2 | 414 | 92 | ||
| XM_015125025.2 | XP_014980511.2 | 446 | 92 | |||
| XM_015125026.2 | XP_014980512.2 | 450 | 92 | |||
| XM_028838787.1 | XP_028694620.1 | 741 | 92 | |||
| XM_028838789.1 | XP_028694622.1 | 507 | 92 | |||
| XM_028838790.1 | XP_028694623.1 | 413 | 92 | |||
| XM_028838791.1 | XP_028694624.1 | 646 | 92 | |||
| GenBank mRNA | JV722891.1 | - | 112 | 92 | ||
More Information
Set Membership: |
|

ZNF135
Genomic Map