Assay Details
| Gene Symbol: CEACAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124085.2 | XP_014979571.2 | 809 | 74 | ||
| XM_028838900.1 | XP_028694733.1 | 802 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM7| Gene Symbol: CEACAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124085.2 | XP_014979571.2 | 809 | 74 | ||
| XM_028838900.1 | XP_028694733.1 | 802 | 74 | |||
Set Membership: |
|