Assay Details
| Gene Symbol: LOC696244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125747.2 | XP_014981233.2 | 187 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015125747.2 transcripts.
Set Membership: |
|

LOC696244
Genomic Map