Assay Details
| Gene Symbol: PFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086728.4 | XP_001086728.1 | 337 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001086728.4 transcripts.
Set Membership: |
|

PFN3
Genomic Map