Assay Details
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119350.2 | XP_014974836.2 | 1547 | 58 | ||
| XM_015119351.2 | XP_014974837.2 | 1549 | 58 | |||
| XM_015119352.2 | XP_014974838.2 | 1288 | 58 | |||
More Information
Set Membership: |
|

TTLL6
Genomic Map