Assay Details
Gene Symbol: LOC100430938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028852010.1 | XP_028707843.1 | 699 | 60 | ||
XM_028852011.1 | XP_028707844.1 | 699 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
