Assay Details
| Gene Symbol: LOC100430938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852010.1 | XP_028707843.1 | 1456 | 116 | ||
| XM_028852011.1 | XP_028707844.1 | 1456 | 116 | |||
More Information
Set Membership: |
|

LOC100430938
Genomic Map