Assay Details
| Gene Symbol: LOC701315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091267.4 | XP_001091267.3 | 631 | 106 | ||
| Gene Symbol: LOC106994543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122152.2 | XP_014977638.2 | 624 | 106 | ||
More Information
Set Membership: |
|

LOC701315
Genomic Map